Merck
Meta-chlorophenylpiperazine enhances leptin sensitivity in diet-induced obese mice.
British journal of pharmacology (2015-03-31)
Chunling Yan, Yongjie Yang, Kenji Saito, Pingwen Xu, Chunmei Wang, Antentor Othrell Hinton, Xiaofeng Yan, Qi Wu, Qingchun Tong, Joel K Elmquist, Makoto Fukuda, Yong Xu
ABSTRAKT
Most forms of human obesity are characterized by impaired leptin sensitivity and, therefore, the effectiveness of anti-obesity leptin therapy in these leptin-resistant obese patients is marginal. Hence, the development of strategies to increase leptin sensitivity is of high priority in the field of obesity research. We first examined the effects of co-administration of leptin and meta-chlorophenylpiperazine (mCPP), an agonist of 5-HT2C and 5-HT1B receptors, on energy balance in leptin-resistant diet-induced obese (DIO) mice. We further assessed leptin-induced phosphorylation of the STAT-3 (pSTAT3) in various brain regions of DIO mice pretreated with mCPP or in mice genetically lacking 5-HT2C receptors. Co-administration of mCPP with leptin had an additive effect on reducing body weight in DIO mice. Furthermore, mCPP pretreatment in DIO mice enhanced leptin-induced pSTAT3 in the arcuate nucleus, the ventromedial hypothalamic nucleus, and the ventral premammillary nucleus. Finally, deletion of 5-HT2C receptors significantly blunted leptin-induced pSTAT3 in these same hypothalamic regions. Our study provides evidence that drugs, which activate 5-HT2C receptors, could function as leptin sensitizers and be used in combination with leptin to provide additional weight loss in DIO.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
Glycine
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O