MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • [Quantitative determination of seven major absorbed volatile constituents in mice brain, liver and blood after intragastric administration of Asari Radix et Rhizoma suspension by headspace-solid phase microextraction-gas chromatography-mass spectrometry].
[Quantitative determination of seven major absorbed volatile constituents in mice brain, liver and blood after intragastric administration of Asari Radix et Rhizoma suspension by headspace-solid phase microextraction-gas chromatography-mass spectrometry].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2016-01-01)
Zi-Wei Zhang, Guang-Xue Liu, De-Mei Xie, Feng Tian, Yi-Kang Jia, Feng Xu, Ming-Ying Shang, Xuan Wang, Shao-Qing Cai
ABSTRACT
A headspace-solid phase microextraction-gas chromatography-mass spectrometry method(HS-SPME-GC-MS) was adopted for the quantitative study of 4-allylanisole, methyl eugenol, 2,3,5-trimethoxytoluene, 3,4,5-trimethoxytoluene, sarisan, 3,5-dimethoxytoluene and safrole in mice brain, liver tissues and blood after intragastric administration of Asari Radix et Rhizoma. A VF-WAXms (30 m×0.25 mm, 0.25 μm film thickness) capillary column and SPME fiber coated with 65 μm polydimethylsiloxane/divinylbenzene (PDMS/DVB) were used. The calibration curves of seven volatile constituents were established to validate the method's stability (RSD<15%), repeatability (RSD<9.5%), accuracy (RSD<22%), relative recovery (87.0%-108%) and extraction recovery (74.9%-102%). The validated HS-SPME-GC-MS assay was applied to determine the concentrations of seven constituents in liver, brain and blood. The detected contents were 0.22,0.14 μg•g⁻¹,0.25 mg•L⁻¹ (4-allylanisole), 1.1, 0.39 μg•g⁻¹, 0.69 mg•L⁻¹ (methyl eugenol), 0.45, 0.13 μg•g⁻¹, 0.54 mg•L⁻¹ (2,3,5-trimethoxytoluene), 0.51, 0.15 μg•g⁻¹, 0.45 mg•L⁻¹ (3,4,5-trimethoxytoluene), 0.48, 0.039 μg•g⁻¹, 0.69 mg•L ⁻¹ (sarisan), 2.2, 1.2 μg•g⁻¹, 1.5 mg•L⁻¹ (3,5-dimethoxytoluene) and 1.3, 0.67 μg•g⁻¹, 1.1 mg•L⁻¹ (safrole) respectively. This HS-SPME-GC-MS method is rapid and convenient, with a small sample size, and applicable for the analysis and determination of volatile constituents in traditional Chinese medicines, which provides scientific data for further studies on effective substances and toxic substances in Asari Radix et Rhizoma.
Product Description
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub