MilliporeSigma
  • Hydrolysis of phosphate monoesters: a biological problem with multiple chemical solutions.
Hydrolysis of phosphate monoesters: a biological problem with multiple chemical solutions.
Trends in biochemical sciences (1992-03-01)
J B Vincent, M W Crowder, B A Averill
ABSTRACT
Formation of phosphate esters by kinases has long been recognized as an important process in biochemistry, but the reverse reaction, hydrolysis of phosphate esters by phosphatases, has attracted less attention. Recent work suggests that phosphatases are as important as kinases in regulatory processes, and that they constitute a diverse group of enzymes that utilize a variety of chemical means to accelerate phosphate ester hydrolysis.
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Pricing and availability is not currently available.