MilliporeSigma
  • MNM and SNM maintain but do not establish achiasmate homolog conjunction during Drosophila male meiosis.
MNM and SNM maintain but do not establish achiasmate homolog conjunction during Drosophila male meiosis.
PLoS genetics (2019-05-29)
Michael Shoujie Sun, Joe Weber, Ariane C Blattner, Soumya Chaurasia, Christian F Lehner
ABSTRACT
The first meiotic division reduces genome ploidy. This requires pairing of homologous chromosomes into bivalents that can be bi-oriented within the spindle during prometaphase I. Thereafter, pairing is abolished during late metaphase I, and univalents are segregated apart onto opposite spindle poles during anaphase I. In contrast to canonical meiosis, homologous chromosome pairing does not include the formation of a synaptonemal complex and of cross-overs in spermatocytes of Drosophila melanogaster. The alternative pairing mode in these cells depends on mnm and snm. These genes are required exclusively in spermatocytes specifically for successful conjunction of chromosomes into bivalents. Available evidence suggests that MNM and SNM might be part of a physical linkage that directly conjoins chromosomes. Here this notion was analyzed further. Temporal variation in delivery of mnm and snm function was realized by combining various transgenes with null mutant backgrounds. The observed phenotypic consequences provide strong evidence that MNM and SNM contribute directly to chromosome linkage. Premature elimination of these proteins results in precocious bivalent splitting. Delayed provision results in partial conjunction defects that are more pronounced in autosomal bivalents compared to the sex chromosome bivalent. Overall, our findings suggest that MNM and SNM cannot re-establish pairing of chromosomes into bivalents if provided after a chromosome-specific time point of no return. When delivered before this time point, they fortify preformed linkages in order to preclude premature bivalent splitting by the disruptive forces that drive chromosome territory formation during spermatocyte maturation and chromosome condensation during entry into meiosis I.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Pricing and availability is not currently available.