MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Necrotic neurons enhance microglial neurotoxicity through induction of glutaminase by a MyD88-dependent pathway.
Necrotic neurons enhance microglial neurotoxicity through induction of glutaminase by a MyD88-dependent pathway.
Journal of neuroinflammation (2008-10-11)
Teresa F Pais, Catarina Figueiredo, Rui Peixoto, Maria H Braz, Sukalyan Chatterjee
ABSTRACT
Microglia are macrophage-like cells that constantly sense the microenvironment within the central nervous system (CNS). In the event of neuronal stress or injury, microglial cells rapidly react and change their phenotype. This response may lead to a deleterious type of microglial activation, which is often associated with neuroinflammation and neurotoxicity in several neuropathological conditions. We investigated the molecular mechanisms underlying triggering of microglial activation by necrotic neuronal damage. Primary cultures of microglia were used to study the effect of necrotic neurons on microglial inflammatory responses and toxicity towards cerebellar granule neurons (CGN). The mouse hippocampal cell line, HT22, was used in this study as the main source of necrotic neurons to stimulate microglia. To identify the signal transduction pathways activated in microglia, primary microglial cultures were obtained from mice deficient in Toll-like receptor (TLR) -2, -4, or in the TLR adapter protein MyD88. Necrotic neurons, but not other necrotic cell types, induced microglial activation which was characterized by up-regulation of: i) MHC class II; ii) co-stimulatory molecules, i.e. CD40 and CD24; iii) beta2 integrin CD11b; iii) pro-inflammatory cytokines, i.e. interleukin 6 (IL-6), IL-12p40 and tumor-necrosis factor (TNF); iv) pro-inflammatory enzymes such as nitric oxide synthase (iNOS, type II NOS), indoleamine 2,3-dioxygenase (IDO) and cyclooxygenase-2 (COX-2) and increased microglial motility. Moreover, microglia-conditioned medium (MCM) obtained from cultures of activated microglia showed increased neurotoxicity mediated through the N-methyl-D-aspartate receptor (NMDAR). The activation of microglia by necrotic neurons was shown to be dependent on the TLR-associated adapter molecule myeloid differentiation primary response gene (MyD88). Furthermore, MyD88 mediated enhanced neurotoxicity by activated microglia through up-regulation of the expression and activity of glutaminase, an enzyme that produces glutamate, which is an NMDAR agonist. These results show that necrotic neurons activate in microglia a MyD88-dependent pathway responsible for a pro-inflammatory response that also leads to increased neurotoxic activity through induction of glutaminase. This finding contributes to better understanding the mechanisms causing increased neuroinflammation and microglial neurotoxicity in a neurodegenerative environment.
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, powder
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine solution, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin human, ≥95% (HPLC), semisynthetic, powder, non-sterile
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Insulin human, meets USP testing specifications
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture