MilliporeSigma
  • Release of alkaline phosphatase from membranes by a phosphatidylinositol-specific phospholipase C.
Release of alkaline phosphatase from membranes by a phosphatidylinositol-specific phospholipase C.
The Biochemical journal (1977-10-01)
M G Low, J B Finean
PMID588258
ABSTRACT
Purified phosphatidylinositol-specific phospholipase C from Staphylococcus aureus released a substantial proportion of the total alkaline phosphatase activity from a wide range of tissues from several mammalian species. Co-purification of the phospholipase C and alkaline phosphatase-releasing activities and the inhibition of both these activities by iso-osmotic salt solutions suggested that the releasing effect was unlikely to be due to a contaminant.
Sigma-Aldrich
Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus, buffered aqueous glycerol solution, ≥1,000 units/mg protein (Lowry)
Pricing and availability is not currently available.
Sigma-Aldrich
Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Pricing and availability is not currently available.