MilliporeSigma
  • The effect of topical tripeptide-copper complex on healing of ischemic open wounds.
The effect of topical tripeptide-copper complex on healing of ischemic open wounds.
Veterinary surgery : VS (2003-12-03)
Sherman O Canapp, James P Farese, Gregory S Schultz, Santosh Gowda, Anthony M Ishak, Steven F Swaim, James Vangilder, Linda Lee-Ambrose, Frank G Martin
ABSTRACT
To evaluate the effects of topical glycyl-L-histidyl-L-lysine tripeptide-copper complex (TCC; Iamin 2% Gel; Procyte Corporation, Redmond, WA) on healing in ischemic open wounds. Experimental study. Twenty-four adult male Sprague-Dawley rats. Rats were divided into 3 groups: topical TCC, topical TCC vehicle (hydroxypropyl-methylcellulose), and no treatment (control). Six-mm-diameter, full-thickness wounds were created within an ischemic bipedicle skin flap on the dorsum of each rat. Each day, for 13 days, wound margins were traced, and the TCC and TCC vehicle groups were treated topically. Tracings were scanned, and wound perimeter and area were calculated. On days 6, 10, and 13, selected wounds were biopsied and analyzed for tumor necrosis factor alpha (TNF-alpha) and matrix metalloproteinases (MMP) 2 and 9. A significant decrease in wound area was seen in the TCC group, but not the vehicle group, when compared with the control group on days 3 to 5, 6 to 9, and 11 to 13 and when TCC was compared with TCC vehicle on days 3 and 9. On day 13, initial wound area had decreased by 64.5% in the TCC group, 45.6% in the vehicle group, and 28.2% in the control group. On days 6, 10, and 13, TCC-treated wounds contained significantly lower concentrations of TNF-alpha and MMP-2 and MMP-9 than control wounds. Topical TCC resulted in accelerated wound healing in ischemic open wounds. Topical TCC is an effective stimulant of healing of ischemic open wounds in rats and may have an application for the treatment of chronic wounds in other species. Clinical evaluation of topical TCC is warranted.
Sigma-Aldrich
Gly-His-Lys acetate salt, ≥97% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture
Pricing and availability is not currently available.