MilliporeSigma
  • Catalytic ceric ammonium nitrate mediated synthesis of 2-deoxy-1-thioglycosides.
Catalytic ceric ammonium nitrate mediated synthesis of 2-deoxy-1-thioglycosides.
Carbohydrate research (2004-09-01)
Somak Paul, Narayanaswamy Jayaraman
ABSTRACT
Synthesis of 2-deoxy-1-thioglycosides from glycals, mediated by catalytic amounts of ceric ammonium nitrate is reported. Apart from the 2-deoxy-1-thioglycosides, formation of the 2,3-unsaturated enose, corresponding to the Ferrier product, is also observed, especially for the glucal substrates. A radical oxocarbenium ion and a thiolate intermediates are most likely to mediate the reaction. Upon synthesis of 2-deoxy-1-thioglycosides, few representative glycosylation reactions with both aglycosyl and glycosyl acceptors were performed and alpha-anomeric 2-deoxy glycosides were obtained exclusively.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, JIS special grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Pricing and availability is not currently available.