MilliporeSigma
  • Surface plasmon resonance imaging measurements of antibody arrays for the multiplexed detection of low molecular weight protein biomarkers.
Surface plasmon resonance imaging measurements of antibody arrays for the multiplexed detection of low molecular weight protein biomarkers.
Analytical chemistry (2006-09-15)
Hye Jin Lee, Dobrin Nedelkov, Robert M Corn
ABSTRACT
This paper describes a simple methodology for the creation of high-density multiplexed antibody arrays on gold surfaces that can be used to detect low molecular weight protein biomarkers with surface plasmon resonance imaging (SPRI). A one-step carbonyldiimidazole (CDI) surface reaction was utilized to attach antibodies onto alkanethiol-modified gold surfaces and characterized with polarization modulation FT-IR reflection absorption spectroscopy. The CDI chemistry was then employed to create an antibody microarray with array element sizes varying from 750 microm down to 200 microm. As a demonstration, a three-component antibody array was employed to detect two clinically important protein biomarkers, beta2-microglobulin (11.8 kDa) and cystatin C (13.4 kDa). SPRI measurements could simultaneously detect both of these small unlabeled proteins with no cross talk at solution concentrations from 300 nM down to 1 nM. In addition, the adsorption strengths of these biomarkers onto an antibody array were measured with SPRI and compared to those obtained from the kinetic analysis of single-channel angle shift SPR measurements.
Sigma-Aldrich
CDI, reagent grade
Pricing and availability is not currently available.
Sigma-Aldrich
CDI, ≥97.0% (T)
Pricing and availability is not currently available.