MilliporeSigma
  • Total synthesis of landomycin A, a potent antitumor angucycline antibiotic.
Total synthesis of landomycin A, a potent antitumor angucycline antibiotic.
Journal of the American Chemical Society (2011-07-26)
Xiaoyu Yang, Boqiao Fu, Biao Yu
ABSTRACT
The first total synthesis of landomycin A, the longest and most potent antitumor angucycline antibiotic, has been achieved in 63 steps and 0.34% overall yield starting from 2,5-dihydroxybenzoic acid, 3,5-dimethylphenol, triacetyl d-glucal, and d-xylose, with a convergent linear sequence of 21 steps.
Sigma-Aldrich
3,5-Dimethylphenol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucal, 96%
Pricing and availability is not currently available.