MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Thermal tolerance and survival of Cronobacter sakazakii in powdered infant formula supplemented with vanillin, ethyl vanillin, and vanillic acid.
Thermal tolerance and survival of Cronobacter sakazakii in powdered infant formula supplemented with vanillin, ethyl vanillin, and vanillic acid.
Journal of food science (2012-08-21)
Gökçe Polat Yemiş, Franco Pagotto, Susan Bach, Pascal Delaquis
ABSTRACT
The thermal tolerance Cronobacter sakazakii was examined in sterile powdered infant formula (PIF) rehydrated at 58 °C in water or apple juice supplemented with vanillin, ethyl vanillin, or vanillic acid. All three compounds decreased thermal tolerance during-rehydration and the lowest decimal reduction time (D-value, 0.19 ± 0.01 min) was measured in PIF rehydrated in apple juice supplemented with 20 mM vanillic acid. At this level of supplementation no C. sakazakii were detected in PIF stored for 48 h at 10 and 24 h at 21 °C subsequent to a sublethal heat treatment. Thermal tolerance during rehydration and survival in reconstituted PIF were influenced by compound type, concentration, and temperature. Supplementation of PIF with vanillin, ethyl vanillin, or vanillic acid could enhance the safety of PIF or other dehydrated foods contaminated with C. sakazakii.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
3-Ethoxy-4-hydroxybenzaldehyde, ReagentPlus®, 99%
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Ethyl vanillin, ≥98%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG