MilliporeSigma
  • Purification and characterization of a novel laccase from the mushroom Pleurotus nebrodensis.
Purification and characterization of a novel laccase from the mushroom Pleurotus nebrodensis.
Acta biochimica Polonica (2012-09-05)
Guo-Ting Tian, Guo-Qing Zhang, He-Xiang Wang, Tzi Bun Ng
ABSTRACT
A novel laccase with a molecular mass of 64 kDa and the N-terminal sequence AIGPDDTINF was isolated from fresh fruiting bodies of the mushroom Pleurotus nebrodensis. The purification protocol comprised ion exchange chromatography on DEAE-cellulose, CM-cellulose, and Q-Sepharose, and gel filtration on Superdex 75. The laccase was adsorbed on DEAE-cellulose and Q-Sepharose, but not on CM-cellulose. It demonstrated an optimal temperature of 70°C. The enzyme activity increased steadily over the temperature range 20°C-70°C. There was only a slight reduction in activity at 80°C. However, all activity disappeared following exposure to 100°C for 10 minutes. The enzyme activity changed only slightly over the pH range 3-5, with the optimum at pH 5, but underwent a precipitous decline when the pH was elevated to 6, and was undetectable at pH 8 and pH 9.
Product Description
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
N,N-Dimethyl-p-phenylenediamine sulfate salt, 98%
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Pyrogallol, JIS special grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethyl-1,4-phenylenediamine oxalate, 98%
Sigma-Aldrich
N,N-Dimethyl-p-phenylenediamine dihydrochloride, suitable for peroxidase test, ≥99.0% (titration)
Sigma-Aldrich
N,N-Dimethyl-p-phenylenediamine, 97%
Millipore
N,N-Dimethyl-p-phenylenediamine dihydrochloride, suitable for microbiology, ≥99.0%