MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibition by capsaicin and its related vanilloids of compound action potentials in frog sciatic nerves.
Inhibition by capsaicin and its related vanilloids of compound action potentials in frog sciatic nerves.
Life sciences (2013-01-29)
Daisuke Tomohiro, Kotaro Mizuta, Tsugumi Fujita, Yukiko Nishikubo, Eiichi Kumamoto
ABSTRACT
Although capsaicin not only activates transient receptor potential vanilloid-1 (TRPV1) channels but also inhibits nerve conduction, the latter action has not yet been fully examined. The purpose of the present study was to know whether various vanilloids have an inhibitory action similar to that of capsaicin and further to compare their actions with that of local anesthetic procaine. Fast-conducting compound action potentials (CAPs) were recorded from frog sciatic nerve fibers by using the air-gap method. Capsaicin reversibly and concentration-dependently reduced the peak amplitude of the CAP. TRPV1 antagonist capsazepine did not affect the capsaicin activity, and powerful TRPV1 agonist resiniferatoxin had no effect on CAPs, indicating no involvement of TRPV1 channels. Capsaicin analogs and other various vanilloids also inhibited CAPs in a concentration-dependent manner. An efficacy sequence of these inhibitions was capsaicin=dihydrocapsaicin>capsiate>eugenol>guaiacol≥zingerone≥vanillin>vanillylamine. Vanillic acid had almost no effect on CAPs; olvanil and curcumin appeared to be effective less than capsaicin. Capsaicin and eugenol were, respectively, ten- and two-fold effective more than procaine in CAP inhibition, while each of guaiacol, zingerone and vanillin was five-fold effective less than procaine. Various vanilloids exhibit CAP inhibition, the extent of which is determined by the property of the side chain bound to the vanillyl group, and some of them are more effective than procaine. These results may serve to unveil molecular mechanisms for capsaicin-induced conduction block and to develop antinociceptive drugs related to capsaicin.
Sigma-Aldrich
Dihydrocapsaicin, from Capsicum sp., ≥85%
Sigma-Aldrich
Vanillylacetone, ≥96%, FG
Sigma-Aldrich
4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Olvanil, powder
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Procaine hydrochloride, ≥97%
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Guaiacol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG