MilliporeSigma
The first pass metabolism of nifedipine in man.
British journal of clinical pharmacology (1984-12-01)
D G Waller, A G Renwick, B S Gruchy, C F George
ABSTRACT
Oral administration of nifedipine (20 and 30 mg tablets) to six volunteers was associated with a bioavailability of 0.43 and the presence of its nitropyridine analogue in the plasma. This metabolite was present in only trace amounts in samples taken from the same volunteers following i.v. administration of nifedipine. The peak plasma concentrations and area under the plasma concentration-time curve suggest that the nitropyridine analogue is a major, first pass, metabolite of nifedipine.
Product Description
Nifedipine impurity A, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Oxidized Nifedipine, powder, ~95% (HPLC)
Pricing and availability is not currently available.