- Psychiatric complications of Erimin abuse.
Psychiatric complications of Erimin abuse.
Singapore medical journal (1989-02-01)
L H Peh, R Mahendran
PMID2595393
ABSTRACT
Nimetazepam (Erimin) is a benzodiazepine which has become a new drug of abuse in Singapore. Three cases presenting with psychiatric complications had been treated as inpatients over a period of one and a half years (1986 to 1987) in Woodbridge Hospital and Tan Tock Seng Hospital. Their clinical presentations which included drug dependency, withdrawal psychosis and transient drug psychosis are described and discussed.
Product Description
Supelco
Nimetazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.