MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Immobilized acclimated biomass-powdered activated carbon for the bioregeneration of granular activated carbon loaded with phenol and o-cresol.
Immobilized acclimated biomass-powdered activated carbon for the bioregeneration of granular activated carbon loaded with phenol and o-cresol.
Bioresource technology (2013-06-26)
Run-Hong Toh, Poh-Eng Lim, Chye-Eng Seng, Rohana Adnan
ABSTRACT
The objectives of the study are to use immobilized acclimated biomass and immobilized biomass-powdered activated carbon (PAC) as a novel approach in the bioregeneration of granular activated carbon (GAC) loaded with phenol and o-cresol, respectively, and to compare the efficiency and rate of the bioregeneration of the phenolic compound-loaded GAC using immobilized and suspended biomasses under varying GAC dosages. Bioregeneration of GAC loaded with phenol and o-cresol, respectively, was conducted in batch system using the sequential adsorption and biodegradation approach. The results showed that the bioregeneration efficiency of GAC loaded with phenol or o-cresol was basically the same irrespective of whether the immobilized or suspended biomass was used. Nonetheless, the duration for bioregeneration was longer under immobilized biomass. The beneficial effect of immobilized PAC-biomass for bioregeneration is the enhancement of the removal rate of the phenolic compounds via adsorption and the shortening of the bioregeneration duration.
Sigma-Aldrich
Phenol stock solution, 100 mg/dL, standard
Sigma-Aldrich
Pandy′s reagent
Sigma-Aldrich
o-Cresol, ≥99%
Supelco
o-Cresol, analytical standard
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%
Sigma-Aldrich
o-Cresol, SAJ first grade, ≥97.0%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)