MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Determination of safranine T in food samples by CTAB sensitised fluorescence quenching method of the derivatives of calix[4]arene.
Determination of safranine T in food samples by CTAB sensitised fluorescence quenching method of the derivatives of calix[4]arene.
Food chemistry (2013-07-23)
Wenjun Wang, Xiashi Zhu, Chaoguo Yan
ABSTRACT
In this paper, the interaction of safranine T (ST) and calix[4]arene (4,10,16,22-tetramethoxylresorcinarene carboxylic acid derivatives (TRCA)) was investigated with fluorescence spectroscopy. The results was shown that the fluorescence intensity of TRCA could be quenched by ST, and the fluorescence quenching (ΔF=FTRCA-FST-TRCA) was sensitised in cetyltrimethyl ammonium bromide (CTAB). Under the optimal conditions, the linear range of calibration curve for the determination of ST was 0.10-4.00 μg/mL. The detection limit and RSD was 0.034 μg/mL, 2.30% (n=3). The quantum yield (Yu) of ST was approximately 2.0 times higher in the presence of CTAB than that in the absence of CTAB. The method has been applied for the determination of ST in food samples with satisfactory results.
Sigma-Aldrich
Gram-Hucker′s stain solution (III)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Safranin O, suitable for microscopy
Sigma-Aldrich
Safranin O, certified by the BSC
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution B