MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Growth of carbon nanofibers using resol-type phenolic resin and cobalt(II) catalyst.
Growth of carbon nanofibers using resol-type phenolic resin and cobalt(II) catalyst.
Journal of nanoscience and nanotechnology (2013-11-20)
Taeyun Kim, Karina Mees, Ho-Seon Park, Monika Willert-Porada, Chang-Seop Lee
ABSTRACT
This study investigated carbon nanofibers (CNFs) grown on reticulated vitreous carbon (RVC) foam through catalytic deposition of ethylene. Before growing the CNFs, Co(II) on the RVC foam was expected to act as a catalyst by deposition. The preparation of the CNFs was a two-step process. The first step was preparing the RVC from polyurethane (PU) foam. Changes in weight over time were evaluated using two kinds of resol. The change in the mass and state of the sample with the change in temperature was studied during the carbonization process. The second step was to prepare the CNFs. An OH group was attached by the oxidation of the RVC foam. A change in the shape and mass of the sample was observed due to a change in nitric acid concentration and oxidation time. Then, cobalt was deposited to grow CNFs on the RVC foam. Hydrolysis helped to deposit the Co(ll) on the RVC foam. The appropriate time and temperature were investigated for the reduction process. In the last step, CNFs were prepared by the introducing ethylene gas. The resulting samples were analyzed using scanning electron microscopy, energy dispersive spectroscopy, N2-sorption, and X-ray photoelectron spectroscopy.
Product Description
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer