MilliporeSigma
  • Efficacy of a novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel against adult and immature stages of the cat flea (Ctenocephalides felis) on cats.
Efficacy of a novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel against adult and immature stages of the cat flea (Ctenocephalides felis) on cats.
Veterinary parasitology (2014-04-08)
Christine Baker, Eric Tielemans, Joseph B Prullage, S Theodore Chester, Martin Knaus, Steffen Rehbein, Josephus J Fourie, David R Young, William R Everett, Joseph K Rosentel
ABSTRACT
The efficacy of a novel topical combination of fipronil 8.3% (w/v), (S)-methoprene 10% (w/v), eprinomectin 0.4% (w/v) and praziquantel 8.3% (w/v) (BROADLINE(®)) was tested against adult and immature stages of Ctenocephalides felis fleas in six studies. For that purpose, fleas from different colonies from North America, Germany and South Africa were used to induce infestations in cats under laboratory conditions. In each study, between 12 and 16 cats were allocated randomly to 2 groups. Cats in Group 1 were not treated and served as controls. Cats in Group 2 were treated once on Day 0 with BROADLINE(®) at the minimum recommended dosage of 0.12 mg/kg body weight. In 4 studies, all animals were infested experimentally with unfed C. felis (100 ± 5) on Days 2 (or 1), 7, 14, 21, 28 and 35. Live fleas were counted 24h post-treatment or infestation. In 2 additional studies, animals were infested at the same frequency with gravid C. felis fleas (100 ± 5) that were fed previously on an untreated host. Forty-eight hours post-infestation, flea eggs were collected, counted and incubated for the evaluation of the reduction of emergence of adults. The combined curative efficacy against adult fleas at 24h after treatment was 94.3% and the combined preventive efficacy values remained greater than 95.9% at 24h after 5 subsequent weekly infestations. In addition, the product reduced dramatically the emergence of new adult fleas for at least 5 weeks (>98.1% for one month and 93.2% at 5 weeks after infestation), demonstrating its efficiency in preventing environmental contamination by immature stages.
Product Description
Supelco
Methoprene, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Praziquantel, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Praziquantel, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Ivermectin
Pricing and availability is not currently available.
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Praziquantel, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Fipronil, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Fipronil, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
Eprinomectin, PESTANAL®, analytical standard, mixture of isomers
Pricing and availability is not currently available.
Praziquantel for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.