MilliporeSigma
  • In vitro release of ketoprofen suppositories using the USP basket and the flow-through cell dissolution methods.
In vitro release of ketoprofen suppositories using the USP basket and the flow-through cell dissolution methods.
Pakistan journal of pharmaceutical sciences (2014-05-09)
José Raúl Medina, Adrián Roberto Padilla, Marcela Hurtado, Alma Rosa Cortés, Adriana Miriam Domínguez-Ramírez
ABSTRACT
In order to study the release characteristics of ketoprofen suppositories under the hydrodynamic environment generated by USP Apparatus 1 and 4, the dissolution profiles of the Mexican reference product (100 mg) were determined. Phosphate buffer pH 8 and 1% sodium lauryl sulfate (SLS) aqueous solutions were proved as dissolution mediums. Baskets were rotated at 100 rpm with USP Apparatus 1 and different flow rates from 16-32 mL/min with USP Apparatus 4 were used. Drug samples were taken and quantified during 60 min by UV analysis at 260 nm. Mean dissolution time (MDT) and dissolution efficiency (DE) were calculated by model-independent methods. Data were also fitted to several kinetic models. Poor dissolution was found in both dissolution mediums when USP basket method was used (< 10% dissolved) while better results were obtained with USP Apparatus 4 when 1% SLS at 24 mL/min was used (43.6% dissolved, MDT of 25.5 min and DE of 25.0%). Kinetics showed a great variability when the USP Apparatus 1 was used, and Gompertz fitted well for data of 1% SLS at 24 mL/min (R(2)(adjusted) > 0.99). The results suggest the need to establish an adequate dissolution method to evaluate the release kinetics of ketoprofen from suppositories.
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Pricing and availability is not currently available.
Supelco
Ketoprofen, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.