MilliporeSigma
  • Flunarizine and cinnarizine-induced parkinsonism: a historical and clinical analysis.
Flunarizine and cinnarizine-induced parkinsonism: a historical and clinical analysis.
Parkinsonism & related disorders (2004-05-04)
Hélio A G Teive, André R Troiano, Francisco M B Germiniani, Lineu C Werneck
ABSTRACT
Drug-Induced Parkinsonism (DIP) represents the second leading cause of Parkinsonism (PK) in several countries. Flunarizine and cinnarizine are some of the most common drugs that cause DIP. This paper reviews the first description of Flunarizine and Cinnarizine-Induced Parkinsonism (FCIP), as well as the subsequent literature, emphasizing epidemiological, clinical and diagnostic aspects. We reviewed the literature on the subject, with special emphasis on the first description and the later definition of the clinical syndrome that results from chronic use of flunarizine and cinnarizine. In 1984, De Melo-Souza reported the first description of flunarizine-induced PK in five patients. Other reports followed on FCIP, emphasizing the clinical features, which are symmetrical parkinsonism, and depression, affecting mainly elderly women. Eighteen years after the original description, FCIP is a recognized condition with specific clinical features, and is the second most common cause of parkinsonism in many countries.
Product Description
Cinnarizine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Cinnarizine, powder
Pricing and availability is not currently available.