MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Development and optimization of curcumin-loaded mannosylated chitosan nanoparticles using response surface methodology in the treatment of visceral leishmaniasis.
Development and optimization of curcumin-loaded mannosylated chitosan nanoparticles using response surface methodology in the treatment of visceral leishmaniasis.
Expert opinion on drug delivery (2014-05-31)
Pramila Chaubey, Ravi R Patel, Brahmeshwar Mishra
ABSTRACT
The study aims at formulation and optimization of macrophage-targeted curcumin-loaded mannosylated chitosan nanoparticles (Cur-MCNPs) of curcumin (CUR) to improve its therapeutic potential in the treatment of visceral leishmaniasis (VL). Response surface methodology (RSM) using central composite design was employed to study the effect of formulation factors on physicochemical-dependent characteristics. Chitosan was coupled with d-mannose, by reductive amination, to prepare a mannosylated chitosan, a conjugate polymer and a subsequent formulation of Cur-MCNPs. Optimized formulation prepared using RSM was evaluated for in vitro release kinetics at physiological pH 7.4 and endosomal macrophage pH 4.5; in vivo pharmacokinetic profile and targeting potential were evaluated by fluorescence microscopy. Optimized Cur-MCNPs exhibited spherical and smooth surface with a mean particle size of 215 nm, polydispersity index of 0.381, zeta potential of + 24.37 mV and % entrapment efficiency of 82.12%. The pharmacokinetic study of optimized Cur-MCNPs showed significant improvement in the value of mean resident time (39.38 h) compared to free CUR solution (0.30 h) (p < 0.05). In vivo uptake study indicated that endocytosis took place effectively within the macrophages of reticuloendothelial system. Thus, Cur-MCNPs could be considered as a promising delivery strategy towards active targeting of CUR to macrophages for the effective treatment of VL.
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Curcumin, primary reference standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Supelco
Curcumin, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)