MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Propofol (Diprivan®) and Intralipid® exacerbate insulin resistance in type-2 diabetic hearts by impairing GLUT4 trafficking.
Propofol (Diprivan®) and Intralipid® exacerbate insulin resistance in type-2 diabetic hearts by impairing GLUT4 trafficking.
Anesthesia and analgesia (2014-12-02)
Phing-How Lou, Eliana Lucchinetti, Liyan Zhang, Andreas Affolter, Manoj Gandhi, Assem Zhakupova, Martin Hersberger, Thorsten Hornemann, Alexander S Clanachan, Michael Zaugg
ABSTRACT
The IV anesthetic, propofol, when administered as fat emulsion-based formulation (Diprivan) promotes insulin resistance, but the direct effects of propofol and its solvent, Intralipid, on cardiac insulin resistance are unknown. Hearts of healthy and type-2 diabetic rats (generated by fructose feeding) were aerobically perfused for 60 minutes with 10 μM propofol in the formulation of Diprivan or an equivalent concentration of its solvent Intralipid (25 μM) ± insulin (100 mU•L). Glucose uptake, glycolysis, and glycogen metabolism were measured using [H]glucose. Activation of Akt, GSK3β, AMPK, ERK1/2, p38MAPK, S6K1, JNK, protein kinase Cθ (PKCθ), and protein kinase CCβII (PKCβII) was determined using immunoblotting. GLUT4 trafficking and phosphorylations of insulin receptor substrate-1 (IRS-1) at Ser307(h312), Ser1100(h1101), and Tyr608(hTyr612) were measured. Mass spectrometry was used to determine acylcarnitines, phospholipids, and sphingolipids. Diprivan and Intralipid reduced insulin-induced glucose uptake and redirected glucose to glycogen stores in diabetic hearts. Reduced glucose uptake was accompanied by lower GLUT4 trafficking to the sarcolemma. Diprivan and Intralipid inactivated GSK3β but activated AMPK and ERK1/2 in diabetic hearts. Only Diprivan increased phosphorylation of Akt(Ser473/Thr308) and translocated PKCθ and PKCβII to the sarcolemma in healthy hearts, whereas it activated S6K1 and p38MAPK and translocated PKCβII in diabetic hearts. Furthermore, only Diprivan phosphorylated IRS-1 at Ser1100(h1101) in healthy and diabetic hearts. JNK expression, phosphorylation of Ser307(h312) of IRS-1, and PKCθ expression and translocation were increased, whereas GLUT4 expression was reduced in insulin-treated diabetic hearts. Phosphatidylglycerol, phosphatidylethanolamine, and C18-sphingolipids accumulated in Diprivan-perfused and Intralipid-perfused diabetic hearts. Propofol and Intralipid promote insulin resistance predominantly in type-2 diabetic hearts.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Soybean oil, CP
Supelco
Soybean oil, analytical standard
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, ≥85% anhydrous basis (enzymatic)
Sigma-Aldrich
Soybean oil, dietary source of long-chain triglycerides and other lipids
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Glucose stock solution, 1000 mg/dL
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Glycogen from bovine liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from oyster, ≥75% dry basis
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
2,6-Diisopropylphenol, 97%