MilliporeSigma
  • Tissue-specific effects of valproic acid on DNA repair genes and apoptosis in postimplantation mouse embryos.
Tissue-specific effects of valproic acid on DNA repair genes and apoptosis in postimplantation mouse embryos.
Toxicological sciences : an official journal of the Society of Toxicology (2014-06-11)
Christina Lamparter, Louise M Winn
ABSTRACT
Exposure to the anticonvulsant drug valproic acid (VPA) is associated with an increased risk of congenital malformations. Although the mechanisms contributing to its teratogenicity are poorly understood, VPA has been shown to induce DNA double strand breaks (DSB) and to increase homologous recombination in vitro. The objective of the present study was to determine whether in utero exposure to VPA alters the frequency of intrachromosomal recombination and the expression of several genes involved in DSB repair in pKZ1 mouse embryos. Pregnant pKZ1 transgenic mice (GD 9.0) were administered VPA (500 mg/kg s.c.) and embryos were extracted and microdissected into the head, heart, and trunk regions 1, 3, 6, and 24 h after injection. Quantitative PCR was used to measure the tissue-specific expression of lacZ, a surrogate measure of recombination, Xrcc4, Rad51, Brca1, and Brca2, with Western blotting used to quantify Rad51, cleaved caspase-3 and cleaved-PARP protein. Increased recombination was only observed in the embryonic head following 6-h VPA exposure. VPA had no effect on Xrcc4 expression. Rad51, Brca1, and Brca2 expression rapidly decreased in head and trunk tissues after 1-h VPA exposure, followed by a subsequent increase in all tissues, although it was generally attenuated in the head and not due to differences in endogenous levels. Cleaved caspase-3 and cleaved-PARP expression was increased in all tissues 3 h following VPA exposure. This study indicates that the tissue-specific expression of several genes involved in DSB repair is altered following exposure to VPA and may be contributing to increased apoptosis.
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2