MilliporeSigma
Electrokinetic bioprocessor for concentrating cells and molecules.
Analytical chemistry (2004-12-02)
Pak Kin Wong, Che-Yang Chen, Tza-Huei Wang, Chih-Ming Ho
ABSTRACT
Bioprocessors for concentrating bioparticles, such as cells and molecules, are commonly needed in bioanalysis systems. In this microfluidic processor, a global flow field generated by ac electroosmosis transports the embedded particles to the regions near the electrode surface. The processor then utilizes electrophoretic and dielectrophoretic forces, which are effective in short range, to trap the target cells and molecules on the electrode surface. By optimizing the operating parameters, we have concentrated various biological objects in a large range of sizes, including Escherichia coli bacteria, lambda phage DNA, and single-stranded DNA fragments as small as 20 bases that have a radius of gyration of only 3 nm.
Sigma-Aldrich
Tris-EDTA buffer solution, 100X concentrate for Northern and Southern blotting
Pricing and availability is not currently available.