MilliporeSigma
  • Peptide combinatorial libraries identify TSC2 as a death-associated protein kinase (DAPK) death domain-binding protein and reveal a stimulatory role for DAPK in mTORC1 signaling.
Peptide combinatorial libraries identify TSC2 as a death-associated protein kinase (DAPK) death domain-binding protein and reveal a stimulatory role for DAPK in mTORC1 signaling.
The Journal of biological chemistry (2008-11-01)
Craig Stevens, Yao Lin, Ben Harrison, Lindsay Burch, Rachel A Ridgway, Owen Sansom, Ted Hupp
ABSTRACT
Death-associated protein kinase (DAPK) is a multidomain enzyme that plays a central role in autophagic and apoptotic signaling, although the protein-protein interactions regulating DAPK functions are not well defined. Peptide aptamer libraries were used to identify the tumor suppressor protein tuberin (TSC2) as a novel DAPK death domain-binding protein, and we evaluated whether DAPK is a positive or negative effector of the TSC2-regulated mammalian target of rapamycin (mTORC1) signaling pathway. Binding studies using death domain miniproteins in vitro and deletion analysis in vivo determined that the death domain of DAPK is the major site for the interaction with TSC2. Recombinant DAPK phosphorylates TSC2 in vitro, and DAPK kinase activity is stimulated by growth factor signaling. Transfection of DAPK promotes phosphorylation of TSC2 in vivo, whereas short interfering RNA-mediated attenuation of DAPK reduces growth factor-stimulated phosphorylation of TSC2. DAPK-dependent phosphorylation leads to TSC1-TSC2 complex dissociation, and consequently manipulation of DAPK by transfection or short interfering RNA demonstrated that DAPK is a positive regulator of mTORC1 in response to growth factor activation. Epistatic studies suggest that DAPK functions downstream from the RAS-MEK-ERK and phosphatidylinositol 3-kinase-AKT growth factor signaling pathways. DAPK(+/-) mouse embryo fibroblasts have attenuated mTORC1 signaling compared with DAPK+/+ counterparts, and overexpression of DAPK in DAPK(+/-) MEFs stimulates mTORC1 activity. These data uncover a novel interaction between DAPK and TSC2 proteins that has revealed a positive link between growth factor stimulation of DAPK and mTORC1 signaling that may ultimately affect autophagy, cell survival, or apoptosis.
Sigma-Aldrich
Streptavidin−Peroxidase from Streptomyces avidinii, lyophilized powder
Pricing and availability is not currently available.