MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • MiR-486 regulates lactation and targets the PTEN gene in cow mammary glands.
MiR-486 regulates lactation and targets the PTEN gene in cow mammary glands.
PloS one (2015-03-05)
Dan Li, Xuejiao Xie, Jie Wang, Yanjie Bian, Qingzhang Li, Xuejun Gao, Chunmei Wang
ABSTRACT
Mammary gland development is controlled by several genes. Although miRNAs have been implicated in mammary gland function, the mechanism by which miR-486 regulates mammary gland development and lactation remains unclear. We investigated miR-486 expression in cow mammary gland using qRT-PCR and ISH and show that miR-486 expression was higher during the high-quality lactation period. We found that miR-486 targets phosphoinositide signaling in the cow mammary gland by directly downregulating PTEN gene expression and by altering the expression of downstream genes that are important for the function of the mammary gland, such as AKT, mTOR. We analyzed the effect of β-casein, lactose and triglyceride secretion in bovine mammary gland epithelial cells (BMECs) transfected by an inhibitor and by mimics of miR-486. Our results identify miR-486 as a downstream regulator of PTEN that is required for the development of the cow mammary gland.
Product Description
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Digoxin, United States Pharmacopeia (USP) Reference Standard
Digoxin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Supelco
Digoxin, analytical standard
Galactose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Lactose, tested according to Ph. Eur.