MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Decrease of Functional Activated T and B Cells and Treatment of Glomerulonephitis in Lupus-Prone Mice Using a Natural Flavonoid Astilbin.
Decrease of Functional Activated T and B Cells and Treatment of Glomerulonephitis in Lupus-Prone Mice Using a Natural Flavonoid Astilbin.
PloS one (2015-04-14)
Lele Guo, Wen Liu, Tingting Lu, Wenjie Guo, Jian Gao, Qiong Luo, Xuefeng Wu, Yang Sun, Xudong Wu, Yan Shen, Qiang Xu
ABSTRACT
Treatment of systemic lupus erythematosus (SLE), a chronic inflammatory disease, involves the long-term use of immunosuppressive agents with significant side effects. New therapeutic approaches are being explored to find better treatment possibilities. In this study, age-matched female MRL/lpr mice were treated orally with a natural flavonoid astilbin. Astilbin administration started either at week 8 or week 12 of age though week 20. In the early treatment regimen, the treatment with astilbin reduced splenomegaly/lymphomegaly, autoantibody production and ameliorated lupus nephritis. Several serum cytokines were significantly decreased upon treatment including IFN-g, IL-17A, IL-1b, TNF-a and IL-6. Both spleen CD44 hi CD62L lo activated T cells and CD138+B220- plasma cells greatly declined. Furthermore, astilbin treatment resulted in decreased mitochondrial membrane potential in activated T cells and downregulated expression of the co-stimulatory molecules CD80 and CD86 on LPS stimulated B cells. Similar but less profound effectiveness was observed in the mice with established disease in the late treatment regimen. These results indicate that the natural product astilbin can mitigate disease development in lupus-prone mice by decreasing functional activated T and B cells.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Resveratrol, European Pharmacopoeia (EP) Reference Standard