MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The antidepressant venlafaxine disrupts brain monoamine levels and neuroendocrine responses to stress in rainbow trout.
The antidepressant venlafaxine disrupts brain monoamine levels and neuroendocrine responses to stress in rainbow trout.
Environmental science & technology (2014-10-31)
Nataliya Melnyk-Lamont, Carol Best, Manuel Gesto, Mathilakath M Vijayan
ABSTRACT
Venlafaxine, a serotonin-norepinephrine reuptake inhibitor, is a widely prescribed antidepressant drug routinely detected in the aquatic environment. However, little is known about its impact on the physiology of nontarget organisms. We tested the hypothesis that venlafaxine perturbs brain monoamine levels and disrupts molecular responses essential for stress coping and feeding activity in fish. Rainbow trout (Oncorhynchus mykiss) were exposed to waterborne venlafaxine (0.2 and 1.0 μg/L) for 7 days. This treatment elevated norepinephrine, serotonin, and dopamine levels in the brain in a region-specific manner. Venlafaxine also increased the transcript levels of genes involved in stress and appetite regulation, including corticotropin releasing factor, pro-opiomelanocortin B, and glucose transporter type 2 in distinct brain regions of trout. The drug treatment reduced the total feed consumed per day, but did not affect the feeding behavior of the dominant and subordinate fish. However, the subordinate fish from the venlafaxine-exposed group had significantly higher plasma cortisol levels compared to the subordinate fish in the control group. Collectively, our results demonstrate that venlafaxine, at environmentally realistic levels, is a neuroendocrine disruptor, impacting the stress and feeding responses in rainbow trout. We propose the midbrain region as a key target for venlafaxine impact and the mode of action involves abnormal monoamine content in trout.
Product Description
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Venlafaxine hydrochloride, ≥98% (HPLC), powder
USP
Venlafaxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Venlafaxine for system suitability, European Pharmacopoeia (EP) Reference Standard
Venlafaxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard