MilliporeSigma
  • Comparison of new point-of-care troponin assay with high sensitivity troponin in diagnosing myocardial infarction.
Comparison of new point-of-care troponin assay with high sensitivity troponin in diagnosing myocardial infarction.
International journal of cardiology (2014-12-17)
Sally Aldous, A Mark Richards, Peter M George, Louise Cullen, William A Parsonage, Dylan Flaws, Christopher M Florkowski, Richard W Troughton, Jack W O'Sullivan, Christopher M Reid, Laura Bannister, Martin Than
ABSTRACT
The aim of this study is to compare a new improved point of care cardiac troponin assay (new POC-cTnI) with 1. its predecessor (old POC-cTnI) and 2. a high sensitivity assay (hs-cTnI) for the diagnosis of acute myocardial infarction (AMI) and for major adverse cardiac events (MACE) by 30 days. This is a single centre observational study, set in Christchurch Hospital, New Zealand. Patients presenting to the emergency department with non-traumatic chest pain underwent blood sampling at 0 h and 2h post presentation for analysis with the 3 cTnI assays for the outcome of AMI and for analysis using an accelerated diagnostic protocol (ADP-normal 2h troponins, normal electrocardiograms and Thrombolysis In Myocardial Infarction (TIMI) score of 0 or ≤ 1) for 30 day MACE. Of 962 patients, 220 (22.9%) had AMI. Old POC-cTnI was least sensitive at 70.0% (65.4-73.9%) by 2h (p<0.001). New POC-cTnI, sensitivity 93.6% (89.9-96.2%) had similar sensitivity to hs-cTnI, sensitivity 95.0% (91.5-97.3%) (p = 0.508). There were 231 (24.0%) patients with 30 day MACE. When used as part of the ADP, all assays had 100% (98.0-100%) sensitivity using TIMI = 0. Sensitivities of new POC-cTnI ADP, 98.3% (95.4-99.4%), old POC-cTnI, 96.5% (93.2-98.4%) and hs-cTnI, 98.7% (96.0-99.7%) were similar (p = 0.063-0.375) using TIMI ≤ 1. A new POC-cTnI has improved sensitivity for AMI and MACE compared with its predecessor and comparable sensitivity to a high sensitivity assay. Now that sensitivities of the POC assay are improved, the new assay may be a useful alternative to central laboratory assays when rapid turn-around times are not possible.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%