MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A comparative study of the effects of inhibitory cytokines on human natural killer cells and the mechanistic features of transforming growth factor-beta.
A comparative study of the effects of inhibitory cytokines on human natural killer cells and the mechanistic features of transforming growth factor-beta.
Cellular immunology (2014-06-01)
Hae Mi Lee, Kyung-Sup Kim, Jongsun Kim
ABSTRACT
The major factors and mechanisms by which natural killer (NK) cells are inhibited in cancer patients have not yet been well defined. In this study, we conducted a comparative analysis of the effects of TGF-β, IL-10, and IL-4 on primary NK cells, and it was demonstrated that (1) TGF-β most potently inhibited the overall function of NK cells. (2) It appears that TGF-β reduced the tyrosine phosphorylation of Syk and the expression of c-myc. (3) It was also found that the IL-2-induced promoter-binding activities of C-myb, AP-1, CREB, and AR were also completely suppressed upon TGF-β treatment. Interestingly, TGF-β also completely suppressed other transcription factors, which are constitutively activated. Among these factors, we further confirmed roles of AP-1 in NK-92 cell activation through c-jun and MEK1 inhibitor assay. Our study provides insight into the effects of TGF-β in modulating NK cell functions.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)