MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Chemoenzymatic synthesis of functional sialyl Lewis(x) mimetics with a heteroaromatic core.
Chemoenzymatic synthesis of functional sialyl Lewis(x) mimetics with a heteroaromatic core.
Chemistry, an Asian journal (2014-06-04)
Claudine Schlemmer, Christine Wiebe, Dorota Ferenc, Danuta Kowalczyk, Stefanie Wedepohl, Patrick Ziegelmüller, Jens Dernedde, Till Opatz
ABSTRACT
Functional mimetics of the sialyl Lewis(X) tetrasaccharide were prepared by the enzymatic sialylation of a 1,3-diglycosylated indole and a glycosyl azide, which was subsequently transformed into a 1,4-diglycosylated 1,2,3-triazole, by using the trans-sialidase of Trypanosoma cruzi. These compounds inhibited the binding of E-, L-, and P-selectin-coated nanoparticles to polyacrylamide-bound sialyl-Lewis(X) -containing neighboring sulfated tyrosine residues (sTyr/sLe(X) -PAA) at low or sub-millimolar concentrations. Except for E-selectin, the mimetics showed higher activities than the natural tetrasaccharide.
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%