MilliporeSigma
  • A mixed MDPV and benzodiazepine intoxication in a chronic drug abuser: determination of MDPV metabolites by LC-HRMS and discussion of the case.
A mixed MDPV and benzodiazepine intoxication in a chronic drug abuser: determination of MDPV metabolites by LC-HRMS and discussion of the case.
Forensic science international (2014-09-10)
Elisabetta Bertol, Francesco Mari, Rafael Boscolo Berto, Guido Mannaioni, Fabio Vaiano, Donata Favretto
ABSTRACT
We report on a case of repeated MDPV consumptions that resulted in severe psychosis and agitation prompting the concomitant abuse of benzodiazepines. A 27-year-old man was found irresponsive in his apartment and was brought to the emergency department (ED) of a local hospital. When in ED, he rapidly recovered and self-reported to have recently injected some doses of MDPV that he had bought in the Internet. He left the hospital without medical cares. 15 days after, he was again admitted to the same ED due to severe agitation, delirium and hallucinations, and reported the use of MDPV and pharmaceutical drugs during the preceding week. He was sedated with diazepam and chlorpromazine. Urine samples collected in both occasions were sent for testing using liquid chromatography-high resolution mass spectrometry (LC-HRMS) and liquid chromatography-high resolution multiple mass spectrometry (LC-HRMS/MS) on an Orbitrap. The LC-HRMS analysis revealed the presence of MDPV and its phase I and phase II metabolites (demethylenyl-MDPV, demethylenyl-methyl-MDPV, demethylenyl-methyl-oxo-MDPV, demethylenyl-hydroxy-alkyl-MDPV, demethylenyl-methyl-hydroxy alkyl-MDPV, demethylenyl-oxo-MDPV and their corresponding glucuronides), alprazolam and alprazolam metabolite at the first ED admission; at the time of the second ED access, the same MDPV metabolites, alprazolam, temazepam, and chlordiazepoxide were detected together with diazepam and metabolites. LC-HRMS/MS was use to determine the following concentrations, respectively on his first and second admission: MDPV 55ng/mL, alprazolam 114ng/mL, α-hydroxyalprazolam 104ng/mL; MDPV 35ng/mL, alprazolam 10.4ng/mL, α -hydroxyalprazolam 13ng/mL; chlordiazepoxide 13ng/mL, temazepam 170ng/mL, diazepam 1.3ng/mL, nordiazepam 61.5, oxazepam 115ng/mL. The toxicological findings corroborated the referred concomitant use of multiple pharmaceutical drugs and benzodiazepines. Confirmation of previous hypothesis on human metabolism of MDPV could be inferred by the analysis of urine.
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Oxazepam
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Nordiazepam solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material