MilliporeSigma
  • Simultaneous analysis of amphetamine-type stimulants in plasma by solid-phase microextraction and gas chromatography-mass spectrometry.
Simultaneous analysis of amphetamine-type stimulants in plasma by solid-phase microextraction and gas chromatography-mass spectrometry.
Journal of analytical toxicology (2014-07-21)
Kristiane de Cássia Mariotti, Roselena S Schuh, Priscila Ferranti, Rafael S Ortiz, Daniele Z Souza, Flavio Pechansky, Pedro E Froehlich, Renata P Limberger
ABSTRACT
Brazil is considered one of the countries with the highest number of amphetamine-type stimulant (ATS) users worldwide, mainly diethylpropion (DIE) and fenproporex (FEN). The use of ATS is mostly linked to diverted prescription stimulants and this misuse is widely associated with (ab)use by drivers. A validated method was developed for the simultaneous analysis of amphetamine (AMP), DIE and FEN in plasma samples employing direct immersion-solid-phase microextraction, and gas chromatographic/mass spectrometric analysis. Trichloroacetic acid 10% was used for plasma deproteinization. In situ derivatization with propylchloroformate was employed. The linear range of the method covered from 5.0 to 100 ng/mL. The detection limits were 1.0 (AMP), 1.5 (DIE) and 2.0 ng/mL (FEN). The accuracy assessment of the control samples was within 85.58-108.33% of the target plasma concentrations. Recoveries ranged from 46.35 to 84.46% and precision was <15% of the value of relative standard deviation. This method is appropriate for screening and confirmation in plasma forensic toxicology analyses of these basic drugs.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard