MilliporeSigma
  • Betahistine ameliorates olanzapine-induced weight gain through modulation of histaminergic, NPY and AMPK pathways.
Betahistine ameliorates olanzapine-induced weight gain through modulation of histaminergic, NPY and AMPK pathways.
Psychoneuroendocrinology (2014-07-06)
Jiamei Lian, Xu-Feng Huang, Nagesh Pai, Chao Deng
ABSTRACT
Olanzapine is widely used to treat schizophrenia and other disorders, but causes adverse obesity and other metabolic side-effects. Both animal and clinical studies have shown that co-treatment with betahistine (a histaminergic H1 receptor agonist and H3 receptor antagonist) is effective for ameliorating olanzapine-induced weight gain/obesity. To reveal the mechanisms underlying these effects, this study investigated the effects of co-treatment of olanzapine and betahistine (O+B) on expressions of histaminergic H1 receptor (H1R), AMP-activated protein kinase (AMPK), neuropeptide Y (NPY), and proopiomelanocortin (POMC) in the hypothalamus associated with reducing olanzapine-induced weight gain. Olanzapine significantly upregulated the mRNA and protein expressions of H1R, while O+B co-treatment significantly downregulated the H1R levels, compared to the olanzapine-only treatment group. The NPY mRNA expression was significantly enhanced by olanzapine, but it was significantly reversed by O+B co-treatment. The hypothalamic H1R expression was positively correlated with total food intake, and NPY expression. Olanzapine also increased AMPKα activation measured by the AMPKα phosphorylation (pAMPKα)/AMPKα ratio compared with controls, whereas O+B co-treatment decreased the pAMPKα/AMPKα ratio, compared with olanzapine only treatment. The pAMPKα/AMPKα ratio was positively correlated with total food intake and H1R expression. Although olanzapine administration decreased the POMC mRNA level, this level was not affected by O+B co-treatment. Therefore, these results suggested that co-treatment with betahistine may reverse olanzapine-induced body weight gain via the H1R-NPY and H1R-pAMPKα pathways.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard