MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Polymeric micelles for pH-responsive delivery of cisplatin.
Journal of drug targeting (2014-06-01)
Mostafa Shahin, Nazila Safaei-Nikouei, Afsaneh Lavasanifar
ABSTRACT
Methoxy poly(ethylene oxide)-block-poly-(α-carboxylate-ε-caprolactone) (PEO-b-PCCL) was used to develop pH-responsive polymeric micelles for the delivery of cisplatin (CDDP). Micelles were prepared through complexation of CDDP with the pendant carboxyl groups on the poly(ε-caprolactone) core, perhaps through coordinate bonding. The obtained micelles were characterized using dynamic light scattering (DLS) measurement for size and stability. The in vitro release of CDDP at different pHs (7.4, 6.0 and 5.0) was evaluated. The in vitro cell uptake as well as cytotoxicity of developed micelles against two breast cancer cell lines, i.e. MDA-MB-435 and MDA-MB-231, were also assessed and compared to free CDDP as control. DLS results showed PEO-b-PCCL to form stable micelles with an average diameter of <50 nm upon complexation with CDDP. Developed polymeric micelles were capable of slowly releasing CDDP in physiological pH. However, CDDP release from polymeric micelles was triggered upon exposure to electrolytes and/or acidic pHs mimicking that of extracellular tumor microenvironment or intracellular organelles. Consistent with the slow release of CDDP from its polymeric micellar formulation, polymeric micellar CDDP exhibited lower cytotoxicity and CDDP intracellular uptake compared to free drug. The results indicate a great potential for the developed formulation in platinum therapy of breast cancer.
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
SAFC
L-Glutamine
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
n-Butyllithium solution, 10.7 M in hexanes
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, suitable for residue analysis