MilliporeSigma
  • Genetic merit for fertility traits in Holstein cows: V. Factors affecting circulating progesterone concentrations.
Genetic merit for fertility traits in Holstein cows: V. Factors affecting circulating progesterone concentrations.
Journal of dairy science (2014-06-24)
S G Moore, S Scully, J A Browne, T Fair, S T Butler
ABSTRACT
This study investigated the factors affecting circulating progesterone (P4) concentrations in cows with similar genetic merit for milk production traits, but with extremes of good (Fert+) or poor (Fert-) genetic merit for fertility traits. Study 1: 28 cows were enrolled in an ovulation synchronization protocol at 61±13 (±standard deviation) days postpartum, and data are presented for 13 Fert+ and 9 Fert- cows that remained in the study. Progesterone concentrations were determined from d 0 to 9 (d 0=estrus) and on d 7, corpus luteum (CL) volume and blood flow area (BFA) were measured by B-mode and Doppler ultrasonography, respectively. Cows were administered PGF2α on d 7 in the p.m. and d 8 in the a.m. to regress the CL, and 2 controlled internal drug release devices were inserted per vaginum on d 8 in the a.m. Liver biopsies were collected on d 9 and hepatic mRNA abundance of genes involved in P4 catabolism was determined. On d 10, the controlled internal drug release inserts were removed and frequent blood samples were collected to measure the rate of decline in circulating P4. The Fert+ cows tended to have greater dry matter intake compared with Fert- cows (+0.79kg of dry matter/d), but similar milk production (29.82kg/d). After synchronized ovulation, the rate of increase in circulating P4 concentrations was greater in Fert+ cows compared with Fert- cows. No effect of genotype on CL volume was detected, but BFA was 42% greater in Fert+ cows compared with Fert- cows. The Fert- cows had greater mRNA abundance of cytochrome P450, family 3, subfamily A (CYP3A) compared with Fert+ cows, but the mRNA abundance of aldo-keto reductase family 1, member C1 (AKR1C1), AKR1C3, AKR1C4, and cytochrome P450, family 2, subfamily C (CYP2C) were similar. The half-life and metabolic clearance rate of P4 were similar in Fert+ cows and Fert- cows. Study 2: 23 cows were enrolled in an ovulation synchronization protocol at 55±7 (±standard deviation) d postpartum, and data are presented for 13 Fert+ and 8 Fert- cows that remained in the study. On d 4, 7, 10, and 13 (d 0=estrus), CL volume and BFA were measured as in study 1. Progesterone concentrations were measured from d 1 to 13. Corpus luteum volume was 41% greater in Fert+ cows compared with Fert- cows but no effect of genotype on BFA was detected. Mean circulating P4 concentrations were 79% greater in Fert+ cows compared with Fert- cows. Milk yield was similar in both genotypes. The results indicate that greater circulating P4 concentrations were primarily due to greater CL P4 synthetic capacity rather than differences in P4 clearance in this lactating cow genetic model of fertility.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Supelco
Ceftiofur hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
SAFC
Tromethamine
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Trometamol, European Pharmacopoeia (EP) Reference Standard
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)