MilliporeSigma
  • Genetic and phenotypic characterization of the heat shock response in Pseudomonas putida.
Genetic and phenotypic characterization of the heat shock response in Pseudomonas putida.
MicrobiologyOpen (2014-10-11)
Fumihiro Ito, Takayuki Tamiya, Iwao Ohtsu, Makoto Fujimura, Fumiyasu Fukumori
ABSTRACT
Molecular chaperones function in various important physiological processes. Null mutants of genes for the molecular chaperone ClpB (Hsp104), and those that encode J-domain proteins (DnaJ, CbpA, and DjlA), which may act as Hsp40 co-chaperones of DnaK (Hsp70), were constructed from Pseudomonas putida KT2442 (KT) to elucidate their roles. The KTΔclpB mutant showed the same heat shock response (HSR) as the wild-type, both in terms of heat-shock protein (Hsp) synthesis (other than ClpB) and in hsp gene expression; however, the mutant was quite sensitive to high temperatures and was unable to disaggregate into thermo-mediated protein aggregates, indicating that ClpB is important for cell survival after heat stress and essential for solubilization of protein aggregates. On the other hand, the KTΔdnaJ mutant was temperature-sensitive, and formed more protein aggregates (especially of high molecular weight) upon heat stress than did KT. P. putida CbpA, a probable Hsp, partially substituted the functions of DnaJ in cell growth and solubilization of thermo-mediated protein aggregates, and might be involved in the HSR which was regulated by a fine-tuning system(s) that could sense subtle changes in the ambient temperature and control the levels of σ(32) activity and quantity, as well as the mRNA levels of hsp genes.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard