MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Curcumin inhibits growth of prostate carcinoma via miR-208-mediated CDKN1A activation.
Curcumin inhibits growth of prostate carcinoma via miR-208-mediated CDKN1A activation.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-06-03)
Hui Guo, Yuemin Xu, Qiang Fu
ABSTRACT
Prostate cancer (PC) is a prevalent cancer in aged men. Curcumin is an active ingredient that has been extracted from the rhizome of the plant Curcuma longa. Recently, a potential of Curcumin against PC has been reported in PC, whereas the underlying molecular mechanisms are not completely understood. Here, we studied the effects of low-dose Curcumin on PC cell growth. Curcumin (from 0.2 to 0.8 μmol/l) dose-dependently inhibited the proliferation of PC cells, without affecting cell apoptosis. Further analyses showed that Curcumin dose-dependently increased a cell cycle suppressor CDKN1A at protein levels, but not mRNA levels, in PC cells, suggesting that Curcumin may regulate the translation of CDKN1A, as well as a possible involvement of miRNA intervention. From all CDKN1A-3'-UTR-binding miRNAs, we found that miR-208 was specifically inhibited in PC cells dose-dependently by Curcumin. Moreover, miR-208 was found to bind CDKN1A to suppress its expression. In a loss-of-function experiment, PC cells that overexpressed miR-208 failed to decrease cell proliferation in response to Curcumin. Together, these data suggest that Curcumin inhibits growth of PC via miR-208-mediated CDKN1A activation.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, meets USP testing specifications