MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Elucidating the Ramularia eucalypti species complex.
Persoonia (2015-08-05)
S I R Videira, J Z Groenewald, A Kolecka, L van Haren, T Boekhout, P W Crous
ABSTRACT
The genus Ramularia includes numerous phytopathogenic species, several of which are economically important. Ramularia eucalypti is currently the only species of this genus known to infect Eucalyptus by causing severe leaf-spotting symptoms on this host. However, several isolates identified as R. eucalypti based on morphology and on nrDNA sequence data of the ITS region have recently been isolated from other plant hosts, from environmental samples and also from human clinical specimens. Identification of closely related species based on morphology is often difficult and the ITS region has previously been shown to be unreliable for species level identification in several genera. In this study we aimed to resolve this species-complex by applying a polyphasic approach involving morphology, multi-gene phylogeny and matrix assisted laser desorption ionization time of flight mass spectrometry (MALDI-TOF MS). Six partial genes (ITS, ACT, TEF1-α, HIS3, GAPDH and RPB2) were amplified and sequenced for a total of 44 isolates representing R. eucalypti s.lat. and closely related species. A multi-gene Bayesian phylogenetic analysis and parsimony analysis were performed, and both the resulting trees showed significant support for separation of seven species in R. eucalypti s.lat., including two previously described (R. eucalypti and R. miae), four novel species here described (R. haroldporteri, R. glennii, R. mali and R. plurivora) and one undescribed Ramularia species (sterile). Additionally, Mycosphaerella nyssicola is newly combined in Ramularia as R. nyssicola. Main mass spectra (MSPs) of several R. eucalypti s.lat. strains were generated using MALDI-TOF MS and were compared through a Principal Component Analysis (PCA) dendogram. The PCA dendrogram supported three clades containing R. plurivora, R. glenni/R. mali and R. eucalypti/R. miae. Although the dendrogram separation of species differed from the phylogenetic analysis, the clinically relevant strains were successfully identified by MALDI-TOF MS.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Perchloric acid, 0.1 M
Sigma-Aldrich
Perchloric acid, SAJ super special grade, 60.0-62.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis