MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Dissipation pattern of flubendiamide residues on capsicum fruit (Capsicum annuum L.) under field and controlled environmental conditions.
Dissipation pattern of flubendiamide residues on capsicum fruit (Capsicum annuum L.) under field and controlled environmental conditions.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2015-11-10)
Radhika Buddidathi, Soudamini Mohapatra, Lekha Siddamallaiah, Gourishankar Manikrao, Shibara Shankara Hebbar
ABSTRACT
This investigation was undertaken to compare the dissipation pattern of flubendiamide in capsicum fruits under poly-house and open field after giving spray applications at the recommended and double doses of 48 g a.i. ha(-1) and 96 g a.i. ha(-1). Extraction and purification of capsicum fruit samples were carried out by the QuEChERS method. Residues of flubendiamide and its metabolite, des-iodo flubendiamide, were analyzed by high-performance liquid chromatography-photodiode array, and confirmed by liquid chromatography-mass spectrometry/mass spectrometry. Limit of quantification of the method was 0.05 mg kg(-1), and recovery of the insecticides was in the range of 89.6-104.3%, with relative standard deviation being 4.5-11.5%. The measurement uncertainty of the analytical method was in the range of 10.7-15.7%. Initial residue deposits of flubendiamide on capsicum fruits grown under poly-house conditions were (0.977 and 1.834 mg kg(-1)) higher than that grown in the field (0.665 and 1.545 mg kg(-1)). Flubendiamide residues persisted for 15 days in field-grown and for 25 days in poly-house-grown capsicum fruits. The residues were degraded with the half-lives of 4.3-4.7 and 5.6-6.6 days in field and poly-house respectively. Des-iodo flubendiamide was not detected in capsicum fruits or soil. The residues of flubendiamide degraded to below the maximum residue limit notified by Codex Alimentarius Commission (FAO/WHO) after 1 and 6 days in open field, and 3 and 10 days in poly-house. The results of the study indicated that flubendiamide applied to capsicum under controlled environmental conditions required longer pre-harvest interval to allow its residues to dissipate to the safe level.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC