MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Evaluation of Acid Digestion Procedures to Estimate Mineral Contents in Materials from Animal Trials.
Evaluation of Acid Digestion Procedures to Estimate Mineral Contents in Materials from Animal Trials.
Asian-Australasian journal of animal sciences (2015-09-04)
M N N Palma, G C Rocha, S C Valadares Filho, E Detmann
ABSTRACT
Rigorously standardized laboratory protocols are essential for meaningful comparison of data from multiple sites. Considering that interactions of minerals with organic matrices may vary depending on the material nature, there could be peculiar demands for each material with respect to digestion procedure. Acid digestion procedures were evaluated using different nitric to perchloric acid ratios and one- or two-step digestion to estimate the concentration of calcium, phosphorus, magnesium, and zinc in samples of carcass, bone, excreta, concentrate, forage, and feces. Six procedures were evaluated: ratio of nitric to perchloric acid at 2:1, 3:1, and 4:1 v/v in a one- or two-step digestion. There were no direct or interaction effects (p>0.01) of nitric to perchloric acid ratio or number of digestion steps on magnesium and zinc contents. Calcium and phosphorus contents presented a significant (p<0.01) interaction between sample type and nitric to perchloric acid ratio. Digestion solution of 2:1 v/v provided greater (p<0.01) recovery of calcium and phosphorus from bone samples than 3:1 and 4:1 v/v ratio. Different acid ratios did not affect (p>0.01) calcium or phosphorus contents in carcass, excreta, concentrate, forage, and feces. Number of digestion steps did not affect mineral content (p>0.01). Estimated concentration of calcium, phosphorus, magnesium, and zinc in carcass, excreta, concentrated, forage, and feces samples can be performed using digestion solution of nitric to perchloric acid 4:1 v/v in a one-step digestion. However, samples of bones demand a stronger digestion solution to analyze the mineral contents, which is represented by an increased proportion of perchloric acid, being recommended a digestion solution of nitric to perchloric acid 2:1 v/v in a one-step digestion.
Sigma-Aldrich
Sodium molybdate, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Perchloric acid, 0.1 M
Sigma-Aldrich
Nitric acid, SAJ first grade, 65.0-66.0%, density: 1.40
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
Nitric acid, 0.1 M
Sigma-Aldrich
Perchloric acid, SAJ super special grade, 60.0-62.0%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%