MilliporeSigma
  • Sphingosine kinase 1 activation enhances epidermal innate immunity through sphingosine-1-phosphate stimulation of cathelicidin production.
Sphingosine kinase 1 activation enhances epidermal innate immunity through sphingosine-1-phosphate stimulation of cathelicidin production.
Journal of dermatological science (2015-06-27)
Se Kyoo Jeong, Young Il Kim, Kyong-Oh Shin, Bong-Woo Kim, Sin Hee Lee, Jeong Eun Jeon, Hyun Jong Kim, Yong-Moon Lee, Theodora M Mauro, Peter M Elias, Yoshikazu Uchida, Kyungho Park
ABSTRACT
The ceramide metabolite, sphingosine-1-phosphate (S1P), regulates multiple cellular functions in keratinocytes (KC). We recently discovered that production of a key innate immune element, cathelicidin antimicrobial peptide (CAMP), is stimulated via a NF-κB-dependent mechanism that is activated by S1P when S1P is generated by sphingosine kinase (SPHK) 1. We investigated whether pharmacological modulation of SPHK1 activity, using a novel synthetic SPHK1 activator, (S)-methyl 2-(hexanamide)-3-(4-hydroxyphenyl) propanoate (MHP), stimulates CAMP expression. MHP-mediated changes in both S1P and CAMP downstream mediators were analyzed in normal cultured human KC by qRT-PCR, Western immunoblot, ELISA, confocal microscopy for immunohistochemistry, HPLC and ESI-LC/MS/MS, and microbial pathogen invasion/colonization in a human epidermal organotypic model. Treatment with MHP directly activated SPHK1 and increased cellular S1P content in normal cultured human KC. Because MHP did not inhibit S1P lyase activity, which hydrolyses S1P, augumented S1P levels could be attributed to increased synthesis rather than blockade of S1P degradation. Next, we found that exogenous MHP significantly stimulated CAMP mRNA and protein production in KC, increases that were significantly suppressed by siRNA directed against SPHK1, but not by a scrambled control siRNA. NF-κB activation, assessed by nuclear translocation of NF-κB, occurred in cells following incubation with MHP. Conversely, pretreatment with a specific inhibitor of SPHK1 decreased MHP-induced nuclear translocation of NF-κB, and significantly attenuated the MHP-mediated increase in CAMP production. Finally, topical MHP significantly suppressed invasion of the virulent Staphylococcus aureus into murine skin explants. MHP activation of SPHK1, a target enzyme of CAMP production, can stimulate innate immunity.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sphk1
SAFC
BIS-TRIS
Sigma-Aldrich
Sphingosine kinase 1 Active human, recombinant, expressed in baculovirus, ≥80% (SDS-PAGE)
Sigma-Aldrich
MISSION® esiRNA, targeting human SPHK1