MilliporeSigma
  • Activity of neurons in the preoptic area and their participation in reproductive senescence: Preliminary findings.
Activity of neurons in the preoptic area and their participation in reproductive senescence: Preliminary findings.
Experimental gerontology (2015-10-13)
Larissa Brazolotto Ferreira, Angela Cristina de Nicola, Janete Aparecida Anselmo-Franci, Rita Cássia Menegati Dornelles
ABSTRACT
Alterations in the hypothalamic-pituitary-gonadal axis in females determine the transition from regular to irregular reproductive cycles, with loss of fertility. Stimulation of noradrenergic neurons of the anteroventral periventricular neurons (AVPV) is essential for regular reproductive cycles. Therefore, we examined the activity of neurons of the AVPV and measure the noradrenaline (NE) of acyclic rats, in constant estrus, and compared it with that of cyclic rats in estrus. Female cyclic (4-5 months) and acyclic (17-18 months) rats were euthanized at 10, 14, and 18 h in estrus. Brains were processed for immunoreactivity to antigens related to Fos (FRA) in AVPV, and the NE was determined by HPLC-ED. Plasma concentrations of LH, FSH, E2 and P4 were determined. In the acyclic animals, plasma LH was higher but the FSH was lower. There was decreasing P4 at different times, while the E2 was constant and lower in acyclic rats. FRA-ir expression in AVPV neurons of acyclic rats as well as turnover of NE was higher when compared with cyclic group. The preliminary findings showed increased activity in AVPV neurons in aging contribute to changes in the temporal pattern of neuroendocrine signaling, compromising the accuracy of inhibitory and stimulatory effects, causing irregularity in the estrous cycle and determining reproductive senescence.
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)