MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Expression of BMI1 and ZEB1 in epithelial-mesenchymal transition of tongue squamous cell carcinoma.
Expression of BMI1 and ZEB1 in epithelial-mesenchymal transition of tongue squamous cell carcinoma.
Oncology reports (2015-06-06)
Kinue Kurihara, Tomohide Isobe, Gou Yamamoto, Yoichi Tanaka, Akira Katakura, Tetsuhiko Tachikawa
ABSTRACT
Epithelial-mesenchymal transition (EMT) is a crucial event required for the invasion and progression of carcinogenesis, inducing stem-like properties in epithelial cells. In the present study, the expression of BMI1, which controls self-renewal in stem cells, as well as that of ZEB1, a transcription factor that regulates EMT, was evaluated for its role in EMT and the carcinogenic processes of tongue squamous cell carcinoma (TSCC). Collagen invasion assays using two TSCC cells and 64 tongue specimens (32 carcinomas and 32 dysplasias) were employed and analyzed in the present study. We assessed the protein and mRNA expression levels of BMI1, ZEB1, vimentin and E-cadherin in the two cell lines and tumor tissues. The protein and mRNA expression of BMI1 and ZEB1 occurred at the invasion of TSCC. The elevated levels of BMI1 and ZEB1 were accompanied by the downregulation of E-cadherin and upregulation of vimentin at the invasive front, indicative of EMT in vitro and in vivo. The results showed that BMI1 and ZEB1 are important factors in association with the promotion of EMT and invasion of TSCC.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O