MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Permeation of steryl ferulates through an in vitro intestinal barrier model.
Permeation of steryl ferulates through an in vitro intestinal barrier model.
Molecular nutrition & food research (2015-03-20)
Dan Zhu, Davide Brambilla, Jean-Christophe Leroux, Laura Nyström
ABSTRACT
Steryl ferulates (SFs) belong to the bioactive lipids contributing to the health promoting effects of whole grains. However, their intestinal absorption remains unclear. We investigated the permeation of individual SFs using an in vitro intestinal barrier model. An in vitro Caco-2 cell monolayer, mimicking the intestinal barrier, was used to evaluate the permeation of eight SFs from different sources. A method based on ultra-performance LC with high-resolution mass spectrometric detection was developed for their quantification. Although only a negligible amount (< 0.5%) permeated across the Caco-2 cell monolayer, some differences in the permeability coefficients were observed between individual SFs. Permeation mechanism was mainly passive diffusion. This work indicates that the permeation of SFs across the gut is very low. Therefore, cholesterol lowering and antioxidant activity-related health benefits of SFs most likely occur in the gut independently from absorption.
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis