MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Proteomic analysis of Corylus heterophylla fisch in Changbai Mountain by shotgun approach.
Proteomic analysis of Corylus heterophylla fisch in Changbai Mountain by shotgun approach.
Pakistan journal of pharmaceutical sciences (2015-10-04)
Liu Chunlei, Min Weihong, Liu Jingsheng, Liu Yan, Ma Jingxi, Li Jingjing
ABSTRACT
Total proteins of Changbai Mountain hazelnut were extracted and then enzymolysed in solution. The mixture of peptides was separated on chromatograph and identified by shotgun proteomics approach. The identified proteins were analyzed by bioinformatics, 303 proteins were identified, of which 237 proteins (78.2%) were mainly distributing in a range of 10-70 kDa and 85 proteins (28.1%) were around pI 5-6. Based on the biological process cellular component and molecular function, these proteins were classified by Gene Ontology, the results showed that 183 proteins (73.8%) had catalytic activity (e.g. oxidoreductase, kinase, peptidase, etc.), and 170 proteins (68.5%) had binding activity. The protein profile provided a valuable basis for further research of hazelnut proteins and opened up new research avenues related to the function of these proteins.
Sigma-Aldrich
Purine, 98%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
3-Indoleacetic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
L-Threonine
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Flavone, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetone, ≥99%, FCC, FG