MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Surface modifications for enhanced enzyme immobilization and improved electron transfer of PQQ-dependent glucose dehydrogenase anodes.
Surface modifications for enhanced enzyme immobilization and improved electron transfer of PQQ-dependent glucose dehydrogenase anodes.
Bioelectrochemistry (Amsterdam, Netherlands) (2015-05-27)
Ryan J Lopez, Sofia Babanova, Kateryna Artyushkova, Plamen Atanassov
ABSTRACT
Pyrroloquinoline quinone dependent soluble glucose dehydrogenase (PQQ-sGDH) enzymatic MWCNT electrodes were p roduced using 1-pyrenecarboxylic acid (PCA) activated through carbodiimide functionalization and 1-Pyrenebutyric acid N-hydroxysuccinimide ester (PBSE) as tethering agents. At 600 mV potential, the current density generated by the activated-PCA tethered PQQ-sGDH anode was significantly greater than the current density generated by the untethered PQQ-sGDH and PBSE tethered anodes, and performance was nearly identical to the performance of a covalently bound PQQ-sGDH anode. A technique for covalently bonding heme-b (hemin), a natural quinohemoprotein porphyrin redox cofactor, to carbon nanotubes modified with arylamine groups is reported. The resulting performance of the covalently bound hemin PQQ-sGDH anode is considerably higher than that of any other PQQ-sGDH anodes tested.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)
Sigma-Aldrich
p-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications