MilliporeSigma
  • Inhibition of autophagy promotes metastasis and glycolysis by inducing ROS in gastric cancer cells.
Inhibition of autophagy promotes metastasis and glycolysis by inducing ROS in gastric cancer cells.
Oncotarget (2015-10-27)
Wenjie Qin, Chao Li, Wen Zheng, Qingqu Guo, Yuefeng Zhang, Muxing Kang, Bo Zhang, Bin Yang, Baozhong Li, Haijun Yang, Yulian Wu
ABSTRACT
Autophagy defect has been shown to be correlated with malignant phenotype and poor prognosis of human cancers, however, the detailed mechanisms remain obscure. In this study, we investigated the biological changes induced by autophagy inhibition in gastric cancer. We showed that inhibition of autophagy in gastric cancer cells promotes epithelial-mesenchymal transition (EMT) and metastasis, alters metabolic phenotype from mitochondrial oxidative phosphorylation to aerobic glycolysis and converts cell phenotype toward malignant, which maybe further contribute to chemoresistance and poor prognosis of gastric cancer. We also identified that the EMT and metabolism alterations induced by autophagy inhibition were dependent on ROS-NF-κB-HIF-1α pathway. More importantly, scavenging of ROS by the antioxidant N-acetylcysteine (NAC) attenuated activation of NF-κB and HIF-1α in autophagy-deficient gastric cancer cells, and autophagy inhibition induced metastasis and glycolysis were also diminished by NAC in vivo. Taken together, our findings suggested that autophagy defect promotes metastasis and glycolysis of gastric cancer, and antioxidants could be used to improve disease outcome for gastric cancer patients with autophagy defect.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-L-cysteine, SAJ special grade, 98.0-102.0%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human RELA
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methoxyestradiol, powder
Pricing and availability is not currently available.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rela
Pricing and availability is not currently available.