MilliporeSigma
  • Production of short-chain fatty acids from the biodegradation of wheat straw lignin by Aspergillus fumigatus.
Production of short-chain fatty acids from the biodegradation of wheat straw lignin by Aspergillus fumigatus.
Bioresource technology (2015-08-12)
Eduardo Baltierra-Trejo, Juan Manuel Sánchez-Yáñez, Otoniel Buenrostro-Delgado, Liliana Márquez-Benavides
ABSTRACT
The wheat straw lignin-rich fraction (WSLig-RF) can be used as a raw material for the production of metabolites for industrial use if ligninolytic mitosporic fungi are used for its biodegradation into aromatics and short-chain fatty acids (SCFAs, i.e., SCFA2-6). Although methods for the production of SCFA2-6 have been described previously, quantitative data of SCFA2-6 production have not been reported. The objectives of this study were to investigate the biodegradation of different concentrations of WSLig-RF by Aspergillus fumigatus and to identify whether SCFA2-6 production was dependent on the concentration of aromatics. A. fumigatus generated 2805mgL(-1) acetic acid when mixed with WSLig-RF at a concentration of 20gL(-1). Thus, aromatics are a substrate for the biosynthesis of SCFA2-6, and their production depends on the concentration of WSLig-RF aromatics.
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Isovaleric acid, ≥99%, FCC, FG
Sigma-Aldrich
Isovaleric acid, 99%
Sigma-Aldrich
Isobutyric acid, ≥99%, FCC, FG